Please login to your online account to display your discounted pricing

Brady™ Glow-In-The-Dark/Ultra Reflective Letter: R

Identify equipment, bins and areas of warehouse required by facility safety and maintenance. Brady™ Glow-In-The-Dark/Ultra Reflective Letter signs reflect light and glow brightly, combining a high level of reflectivity with advanced glow-in-the-dark technology.

$15.72 - $15.72

Specifications

Material Vinyl (B-309)
Color Black
Color Glow
Color Green
Color Silver
View More Specs
Products
Catalog Number Mfr. No. Height (English) Character Height (Metric) Character Size (English) Size (Metric) Price Quantity    

19-108-436

 
brady™
BRD3010R
3 in. Pack of 10 Each for $15.72
Description & Specifications

Specifications

Material Vinyl (B-309)
Color Black
Color Glow
Color Green
Color Silver
Product Type Ultra Reflective Letter: R
For Use With (Application) Labeling in Low Light Conditions (Indoor/Outdoor)
Display Text Legend: R

  • Micro-prism reflectors for maximum visibility from greater distance, wider angle and lower light source
  • Designed for permanent Indoor/Outdoor applications
  • Made of high intensity prismatic reflective sheeting photoluminescent material with pressure sensitive adhesive