Purus™ Cut Resistant Glove Liner
Cut Resistant Glove Liners provide ANSI Cut Level 2 protection. Made from 13 Gauge White Nylon with HPPE, these glove liners provide light cut protection while wicking away moisture during extended use. Color coded cuff indicate size and are sturdy, launderable and reusable. Compatible with cleanroom requirements.
Manufacturer:  Purus™ GLFFXLCR
 View more versions of this product
Catalog No. 19126903
Description
Description
  • Ambidexterous
  • Available in sizes S-XL
  • 100% Quality Control Inspection
  • ANSI Cut Level 2 Protection: Made from 13 Gauge White Nylon with HPPE
  • Compatible underglove for all cleanroom environments
  • Increased worker safety and satisfaction
  • Color Coded Cuffs for visual size identification
  • Sturdy
  • Launderable and Reusable.
Specifications
Specifications
2
13 Gauge White Nylon with HPPE
Full finger
Latex, nitrile, PVC and all exam gloves
Yellow
Compatible with stringent cleanroom requirements
Nylon, HPPE
Extra large
Glove
Powder-free
Non-sterile
Launderable
No
knit wrist
Ambidextrous
Handling of glassware or sharp objects in wet or dry environments
Yes
20 pairs/bag; 10 bags/case (200 pairs/case)
20 pairs/bag; 10 bags/case (200 pairs/case)
Cut Resistant Glove Liner
White
Red
Documents