Please login to your online account to display your discounted pricing

Alfa Aesar™ [Gln&superscript_4;] Neurotensin

For cellular and molecular biology applications

Specifications

Product Type [Gln4] Neurotensin
Molecular Weight 1671.96
Storage Requirements -30° to -10°C
Sequence Pyr-Leu-Tyr-Gln-Asn-Lys-Pro-Arg-Arg-Pro-Tyr-Ile-Leu
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66142LB0

 
alfa aesar™
J66142LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Gln4] Neurotensin
Molecular Weight 1671.96
Storage Requirements -30° to -10°C
Sequence Pyr-Leu-Tyr-Gln-Asn-Lys-Pro-Arg-Arg-Pro-Tyr-Ile-Leu
Format Solid
Common Name Large Neuromedin N
Regulatory Status RUO
Purification Method HPLC