Please login to your online account to display your discounted pricing

Alfa Aesar™ [Gln&superscript_1;&superscript_1;] Amyloid beta (1-16)

For cellular and molecular biology applications

$486.69 - $676.54

Specifications

Product Type [Gln11] Amyloid beta (1-16)
Molecular Weight 1954.02
Storage Requirements -30° to -10°C
Format Solid
Sequence H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Gln-Val-His-His-Gln-Lys-OH
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66868LB0

 
alfa aesar™
J66868LB0
5mg Each for $486.69

AAJ66868MA

 
alfa aesar™
J66868MA
10mg Each for $676.54
Description & Specifications

Specifications

Product Type [Gln11] Amyloid beta (1-16)
Molecular Weight 1954.02
Storage Requirements -30° to -10°C
Format Solid
Sequence H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Gln-Val-His-His-Gln-Lys-OH
Regulatory Status RUO
Purification Method HPLC