Please login to your online account to display your discounted pricing

Alfa Aesar™ [Gln&superscript_1;&superscript_1;] Amyloid beta (1-16)


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66868LB0

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66868LB0

  • / Each
 

Description & Specifications

Specifications

Storage Requirements -30° to -10°C
Sequence H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Gln-Val-His-His-Gln-Lys-OH
Regulatory Status RUO
Quantity 5mg
Format Solid
Molecular Weight 1954.02
Purification Method HPLC
Product Type [Gln11] Amyloid beta (1-16)