Please login to your online account to display your discounted pricing

Glimepiride, Tocris

Kir6 (KATP) channel blocker

$75.35 - $257.85

Chemical Identifiers

Chemical Name or Material Glimepiride
Assay Percent Range >98%
Molecular Formula C24H34N4O5S
Formula Weight 490.62
Synonym 3-Ethyl-2, 5-dihydro-4-methyl-N-[2-[4-[[[[(trans-4-methylcyclohexyl)amino]carbonyl]amino]sulfonyl]phenyl]ethyl]-2-oxo-1H-pyrrole-1-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

239610

 
r & d systems
RDS239610
10mg Each for $75.35

239650

 
r & d systems
RDS239650
50mg Each for $257.85
Description & Specifications

Specifications

Formula Weight 490.62
Quantity 10mg
Chemical Name or Material Glimepiride
Molecular Formula C24H34N4O5S
Assay Percent Range >98%
CAS 93479-97-1
Synonym 3-Ethyl-2, 5-dihydro-4-methyl-N-[2-[4-[[[[(trans-4-methylcyclohexyl)amino]carbonyl]amino]sulfonyl]phenyl]ethyl]-2-oxo-1H-pyrrole-1-carboxamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C