Please login to your online account to display your discounted pricing

Alfa Aesar™ Glassy Carbon Tapered Crucible for Crystal Growth

Glassy carbon offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.

Specifications

Product Type Glassy Carbon Crucible
Material Glassy Carbon
Item Description Tapered
For Use With (Application) For Crystal Growth
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Height (Metric) Item Description Price Quantity    

AA40954KT

 
alfa aesar™
40954KT
18mL Top: 24mm 87mm Tapered CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40956KT

 
alfa aesar™
40956KT
105mL Top: 59mm 59mm Tapered CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40957KT

 
alfa aesar™
40957KT
190mL Top: 45mm 168mm Tapered CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40958KT

 
alfa aesar™
40958KT
277mL Top: 73mm 85mm Tapered CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40959KT

 
alfa aesar™
40959KT
370mL Top: 69mm 133mm Tapered CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Glassy Carbon Crucible
Material Glassy Carbon
Item Description Tapered
For Use With (Application) For Crystal Growth

Subjected to a high temperature heat treatment which imparts special material properties including significantly improved corrosion resistance and strength.

Glassy carbon offers:

  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity