Please login to your online account to display your discounted pricing
Alfa Aesar™ Glassy Carbon Cylindrical Crucible
Glassy carbon offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.
Specifications
Product Type Glassy Carbon Crucible
Material Glassy Carbon
Item Description Cylindrical
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Height (Metric) Item Description Price Quantity    
AA39008KT
alfa aesar™
39008KT
10mL 24mm 39mm Cylindrical CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA39010KT
alfa aesar™
39010KT
30mL 36mm 45mm Cylindrical CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA39011KT
alfa aesar™
39011KT
50mL 40mm 56mm Cylindrical CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA39012KT
alfa aesar™
39012KT
130mL 56mm 85mm Cylindrical CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA39014KT
alfa aesar™
39014KT
260mL 73mm 85mm Cylindrical CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Product Type Glassy Carbon Crucible
Material Glassy Carbon
Item Description Cylindrical
Subjected to a high temperature heat treatment which imparts special material properties including significantly improved corrosion resistance and strength.
Glassy carbon offers:
  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity