Please login to your online account to display your discounted pricing

Alfa Aesar™ Glassy Carbon Crucible for Crystal Growth

Glassy carbon offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.

Specifications

Product Type Glassy Carbon Crucible
Material Glassy Carbon
Item Description Bottom Angle: 90 deg.
For Use With (Application) For Crystal Growth
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Height (Metric) Item Description Price Quantity    

AA39015KT

 
alfa aesar™
39015KT
7mL Top: 14mm; Bottom: 14mm 100mm Bottom Angle: 90 deg. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA39016KT

 
alfa aesar™
39016KT
25mL Top: 19mm; Bottom: 19mm 140mm Bottom Angle: 60 deg. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA39017KT

 
alfa aesar™
39017KT
30mL Top: 24mm; Bottom: 19mm 160mm Bottom Angle: 30 deg. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA39019KT

 
alfa aesar™
39019KT
400mL Top: 57mm; Bottom: 57mm 195mm Bottom Angle: 90 deg. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Glassy Carbon Crucible
Material Glassy Carbon
Item Description Bottom Angle: 90 deg.
For Use With (Application) For Crystal Growth

Subjected to a high temperature heat treatment which imparts special material properties including significantly improved corrosion resistance and strength.

Glassy carbon offers:

  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity