Please login to your online account to display your discounted pricing

Alfa Aesar™ Glassclad™ 18

Chemical Identifiers

Chemical Name or Material Glassclad™ 18
MDL Number MFCD00240276
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA432935Y

 
alfa aesar™
432935Y
4 x 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA43293AC

 
alfa aesar™
43293AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA43293K4

 
alfa aesar™
43293K4
2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA43293AE

 
alfa aesar™
43293AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Glassclad™ 18
Flash Point 10°C (50°F)
Physical Form Liquid
Density 0.90g/mL
MDL Number MFCD00240276
UN Number UN1993
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H332-H319
Highly flammable liquid and vapor.
Harmful if inhaled.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off imm
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Quantity 4 x 100mL

  • As organosilane concentrate which, when properly applied to glass, provides a surface which is water repellent, has improved lubricity, and surface resistivity

Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off imm

GHS H Statement
H225-H332-H319
Highly flammable liquid and vapor.
Harmful if inhaled.
Causes serious eye irritation.