DWK Life Sciences Wheaton™ Glass Sample Vials in Vial File™: PROMO
Vial File provides a compact, easily accessible means of sample storage
Manufacturer:  DWK Life Sciences WW224694PM
 View more versions of this product
Catalog No. 02-912-510PM
Description
Description
Caps attached to vials.
  • Polystyrene case with partitions, foam inserts and alphanumeric indexing card helps organize and protect vials
  • Choice of solid- or open-top black phenolic caps: open caps facilitate sample retrieval with syringe
  • Vials and caps autoclavable
Clear or Amber Vials
  • Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
  • Amber vials made from borosilicate glass that protects light-sensitive products
Compliance
Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements. Amber vials conform to USP Type 1 requirements of light transmission.
Specifications
Specifications
Yes
Screw cap
15mm
Low Extractable Borosilicate Glass
4mL
48mm
Used for storing and transporting vials
Screw
USP Type I, ASTM E438, Type I, Class A
4.0mL
Clear
Glass Sample Vial
No
Phenolic
PTFE/14B Rubber
1.08 dr.