Please login to your online account to display your discounted pricing

Qorpak™ Glass Bottle Beakers with White PP Sturdeeseal™ PE Foam Caps

For mixing, measuring and storage

$87.03 - $711.43

Specifications

Closure Liner SturdeeSeal™ Polyethylene Foam
Closure Material Polypropylene
Closure Screw
Closure Color White
Color Clear
View More Specs
Products
Catalog Number Mfr. No. Capacity Height (English) Quantity Height (Metric) Price Quantity    

02991224

 
qorpak™
GLC05090
4 oz., 120mL 4.02 in. 144/Cs. 102mm Case for $402.91

02991212

 
qorpak™
GLC01578
16 oz., 480mL 5.71 in. 48/Cs. 145mm Case for $235.11

02991335

 
qorpak™
GLC05092
2 oz., 60mL 3.27 in. 288/Cs. 83mm Case for $711.43

02991346

 
qorpak™
GLC01555
8 oz., 240mL 5 in. 96/Cs. 127mm Case for $366.31

02992109

 
qorpak™
GLC06615
16 oz., 480mL 5.71 in. 24/Cs. 145mm Case for $141.43

02992116

 
qorpak™
GLC05089
4 oz., 120mL 4.02 in. 24/Cs. 102mm Case for $87.03

02992130

 
qorpak™
GLC06616
2 oz., 60mL 3.27 in. 48/Cs. 83mm Case for $141.46

02992178

 
qorpak™
GLC06377
8 oz., 240mL 5 in. 24/Cs. 127mm Case for $113.60
Description & Specifications

Specifications

Closure Liner SturdeeSeal™ Polyethylene Foam
Closure Material Polypropylene
Closure Screw
Closure Color White
Color Clear
Material Glass

  • Designed with slight shoulder for easy pouring and handling
  • Molded graduations in mL and ounces
  • Made of clear glass
  • For use with biological and pathological specimens
  • For storage of industrial laboratory chemicals and reagents