missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Qorpak™ Glass Bottle Beakers with White PP Sturdeeseal™ PE Foam Caps
For mixing, measuring and storage
Supplier: Qorpak™ GLC06377
Description
- Designed with slight shoulder for easy pouring and handling
- Molded graduations in mL and ounces
- Made of clear glass
- For use with biological and pathological specimens
- For storage of industrial laboratory chemicals and reagents
Specifications
Polypropylene | |
62 mm | |
Screw Cap | |
58-400 mm | |
240 mL | |
Clear | |
Glass | |
White |
127 mm | |
5 in. | |
Polyethylene Foam | |
8 oz., 240 mL | |
8 oz. | |
2.44 in. | |
24/Cs. |