Please login to your online account to display your discounted pricing

Qorpak™ Glass Bottle Beakers with Green Thermoset F217 and PTFE Caps

Can be used for mixing, measuring and storage

$167.60 - $1,711.77

Specifications

Closure Liner F217 and PTFE
Closure Material Thermoset
Closure Screw
Closure Color Green
Color Clear
View More Specs
Products
Catalog Number Mfr. No. Capacity Height (English) Quantity Height (Metric) Price Quantity    

02992128

 
qorpak™
GLC01450*
1 oz., 30mL 2.68 in. 48/Cs. 68mm Case for $205.43

02992601

 
qorpak™
GLC01472
2 oz., 60mL 3.27 in. 48/Cs. 83mm Case for $256.83

02992588

 
qorpak™
GLC01504*
4 oz., 120mL 4.02 in. 24/Cs. 102mm Case for $167.60

02992647

 
qorpak™
GLC01532
8 oz., 240mL 5 in. 24/Cs. 127mm Case for $217.43

02992801

 
qorpak™
GLC01560
16 oz., 480mL 5.71 in. 24/Cs. 145mm Case for $298.83

02993059

 
qorpak™
GLC01449
1 oz., 30mL 2.68 in. 432/Cs. 68mm Case for $1,711.77

02993085

 
qorpak™
GLC01470
2 oz., 60mL 3.27 in. 288/Cs. 83mm Case for $1,412.09

02993162

 
qorpak™
GLC01531
8 oz., 240mL 5 in. 96/Cs. 127mm Case for $802.83

02993200

 
qorpak™
GLC01502
4 oz., 120mL 4.02 in. 144/Cs. 102mm Case for $874.20

02993218

 
qorpak™
GLC01559
16 oz., 480mL 5.71 in. 48/Cs. 145mm Case for $571.69
Description & Specifications

Specifications

Closure Liner F217 and PTFE
Closure Material Thermoset
Closure Screw
Closure Color Green
Color Clear
Material Glass

  • Designed with slight shoulder for easy pouring and handling
  • Molded graduations in mL and ounces
  • Made of clear glass
  • For use with biological and pathological specimens
  • For storage of industrial laboratory chemicals and reagents