missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GL7 (T and B Cell Activation Marker) Rat anti-Mouse, PE, Clone: GL-7 (GL7), eBioscience™

Rat Monoclonal Antibody

$138.60 - $293.04

Specifications

Antigen GL7 (T and B Cell Activation Marker)
Clone GL-7 (GL7)
Host Species Rat
Gene Alias Ly-77, Ly77
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122343 Life Technologies
LS12590280
25μg Each for $138.60
501129506 Life Technologies
LS12590282
100μg Each for $293.04
Specifications

Description

This GL7 monoclonal antibody reacts with a cell-surface protein found on T and B lymphocytes activated in vitro, on bone marrow pre-B-II cells, germinal center B cells, and also human B cell lines Ramos and Daudi. There is strain variability with respect to antigen distribution on thymocytes and Con A-activated spleen cells, with expression in BALB/c greater than that in C57BL/6. GL7 is commonly used as a marker for mouse germinal center B cells. The epitope of GL7 has been identified as a sialic acid glycan moiety called Neu5Ac. This moiety is recognized by CD22.
Description & Specifications

Specifications

GL7 (T and B Cell Activation Marker)
Rat
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
GL-7 (GL7)
Ly-77, Ly77
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer