missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
GJB5, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00002709B02P
Description
Sequence: MNWSIFEGLLSGVNKYSTAFGRIWLSLVFIFRVLVYLVTAERVWSDDHKDFDCNTRQPGCSNVCFDEFFPVSHVRLWALQLILVTCPSLLVVMHVAYREVQEKRHREAHGENSGRLYLNPGKKRGGLWWTYVCSLVFKASVDIAFLYVFHSFYPKYILPPVVKCHADPCPNIVDCFISKPSEKNIFTLFMVATAAICILLNLVELIYLVSKRCHECLAARKAQAMCTGHHPHGTTSSCKQDDLLSGDLIFLGSDSHPPLLPDRPRDHVKKTILSpecifications
GJB5 | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
NM_005268.2 | |
GJB5 | |
GJB5 (NP_005259.1, 1 a.a. ∼ 273 a.a) full-length human protein. | |
RUO | |
2709 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
gap junction protein, beta 5, 31.1kDa | |
CX31.1 | |
Mouse | |
50 μg | |
Primary | |
Human | |
Antibody |