Please login to your online account to display your discounted pricing

Ginkgolide B, Tocris

PAF receptor antagonist

$113.57 - $478.51

Chemical Identifiers

Chemical Name or Material Ginkgolide B
Assay Percent Range >98%
Molecular Formula C20H24O10
Formula Weight 424.4
Synonym (1R,3S,3aS,4R,6aR,7aR,7bR,8S,10aS,11R,11aR)-3-(1,1-Dimethylethyl)hexahydro-4,7b,11-trihydroxy-8-methyl-9H-1,7a-(epoxymethano)-1H,6aH-cyclopenta[c]furo[2,3-b]furo[3',2':3,4]cyclopenta[1,2-d]furan-5,9,12(4H)-trione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

165750

 
r & d systems
RDS165750
50mg Each for $478.51

165710

 
r & d systems
RDS165710
10mg Each for $113.57
Description & Specifications

Specifications

Formula Weight 424.4
Quantity 50mg
Chemical Name or Material Ginkgolide B
Molecular Formula C20H24O10
CAS 15291-77-7
Recommended Storage Desiccate at +4°C
Assay Percent Range >98%
Synonym (1R,3S,3aS,4R,6aR,7aR,7bR,8S,10aS,11R,11aR)-3-(1,1-Dimethylethyl)hexahydro-4,7b,11-trihydroxy-8-methyl-9H-1,7a-(epoxymethano)-1H,6aH-cyclopenta[c]furo[2,3-b]furo[3',2':3,4]cyclopenta[1,2-d]furan-5,9,12(4H)-trione
Safety and Handling

Desiccate at +4°C