missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ginkgolide B, Thermo Scientific Chemicals

Platelet-Activating Factor (PAF) antagonist

Supplier:  Thermo Scientific Chemicals J60646MA

 View more versions of this product

Catalog No. AAJ60646MA



Description

Description

Ginkgolide B is used as a potent PAF-R antagonist. Ginkgolide B speeds up PAF hydrolysis by promoting PAF-AH I α2 homodimer activity. Additionally, this compound has been reported to inhibit neutrophil degranulation in vitro. Inhibits superoxide production in vitro and inhibits bronchoconstriction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ginkgolide B is used as a potent PAF-R antagonist. Ginkgolide B speeds up PAF hydrolysis by promoting PAF-AH I α2 homodimer activity. Additionally, this compound has been reported to inhibit neutrophil degranulation in vitro. Inhibits superoxide production in vitro and inhibits bronchoconstriction.

Solubility
Soluble in DMSO. Slightly soluble in water and ethanol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away fom strong oxidizing agents.
Specifications

Specifications

Ginkgolide B
White
MFCD05662347
14,4426
Soluble in DMSO; Slightly soluble in water and ethanol
CC1C(=O)OC2C1(C34C(=O)OC5C3(C2O)C6(C(C5)C(C)(C)C)C(C(=O)OC6O4)O)O
132470872
Powder
15291-77-7
C20H24O10
10 mg
sqojoafxdqdrgf-maggbrlhsa-n
SQOJOAFXDQDRGF-WVXYHUDCSA-N
424.402
424.4

Chemical Identifiers

15291-77-7
424.402
SQOJOAFXDQDRGF-WVXYHUDCSA-N
132470872
C20H24O10
MFCD05662347
sqojoafxdqdrgf-maggbrlhsa-n
CC1C(=O)OC2C1(C34C(=O)OC5C3(C2O)C6(C(C5)C(C)(C)C)C(C(=O)OC6O4)O)O
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only