Please login to your online account to display your discounted pricing

Alfa Aesar™ Ginkgolide A

Platelet-Activating Factor (PAF) antagonist

Chemical Identifiers

Chemical Name or Material Ginkgolide A
CAS 15291-75-5
Formula Weight 408.41
MDL Number MFCD00133365
Merck Index 14,4426
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ62384FPL

 
alfa aesar™
J62384FPL
20mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 408.41
MDL Number MFCD00133365
CAS 15291-75-5
Physical Form Powder
Solubility Information Soluble in DMSO
Color White
RTECS Number KC9943000
Quantity 20mg
Chemical Name or Material Ginkgolide A
Merck Index 14,4426
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C