Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ TOWERPACK™ Refill System


Differentiates tip models with color-coded racks and indicates the compatible Gilson pipet models. Gilson™ PIPETMAN™ TOWERPACK™ Refill System is available in autoclavable and sterilized versions.

Manufacturer: gilson™  F167203

 View more versions of this product

Catalog No. F167203G

  • / Pack of 960 Each
 

Description & Specifications

Specifications

Capacity (Metric) 2 - 200μL
For Use With (Equipment) D200 2 to 200μL 51mm L Pipetman™ Diamond™ pipet tips
Sterile Yes
Product Type Tower Pack™ Pipet Refill System

  • Color-coded racks to differentiate tip models and indicate the compatible Gilson™ pipet models
  • Available in autoclavable and sterilized versions
  • Fits Universal Reload box (Cat. No. F167100G, available separately)
  • Sterilized Tower Pack is individually wrapped in medical device-quality plastic to help protect tips from contamination