Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ TOWERPACK™ Refill System


Differentiates tip models with color-coded racks and indicates the compatible Gilson pipet models. Gilson™ PIPETMAN™ TOWERPACK™ Refill System is available in autoclavable and sterilized versions.

Manufacturer: gilson™  F167101

 View more versions of this product

Catalog No. F167101G

  • / Pack of 960 Each
 

Description & Specifications

Specifications

Product Type Tower Pack™ Pipet Refill System
Sterile No
For Use With (Equipment) D10 0.1 to 10μL 30mm L Pipetman™ Diamond™ pipet tips
Capacity (Metric) 0.1 - 10μL

  • Color-coded racks to differentiate tip models and indicate the compatible Gilson™ pipet models
  • Available in autoclavable and sterilized versions
  • Fits Universal Reload box (Cat. No. F167100G, available separately)
  • Sterilized Tower Pack is individually wrapped in medical device-quality plastic to help protect tips from contamination