Please login to your online account to display your discounted pricing
Gilson™ PIPETMAN™ Tipack™ Racked Pipet Tips
Engineered to be functional, convenient and robust. Gilson™ PIPETMAN™ Tipack™ Racked Pipet Tips are available in sterilized or autoclavable packages.
Manufacturer: gilson™  F161230
 View more versions of this product
Catalog No. F161230G
  • / Pack of 240 Each
Description & Specifications
Specifications
Sterility Sterile
Material Polypropylene
Length (Metric) 1.6cm
Model D10mL
Autoclavable Yes
Packaging 10 Tipacks of 96 racked tips
Volume (Metric) 1 to 10mL