Gilson™ PIPETMAN™ Tipack™ Racked Pipet Tips
Engineered to be functional, convenient and robust. Gilson™ PIPETMAN™ Tipack™ Racked Pipet Tips are available in sterilized or autoclavable packages.
Manufacturer:  Gilson™ F171501
 View more versions of this product
Catalog No. F171501G
Specifications Description & Specifications
Specifications
100 to 1000μL
Yes
Polypropylene
5.1cm
Sterile
D1000
10 Tipacks of 96 Racked Tips