Please login to your online account to display your discounted pricing
Gilson™ PIPETMAN™ Tipack™ Racked Pipet Tips
Engineered to be functional, convenient and robust. Gilson™ PIPETMAN™ Tipack™ Racked Pipet Tips are available in sterilized or autoclavable packages.
Manufacturer: gilson™  F171300
 View more versions of this product
Catalog No. F171300G
  • / Pack of 960 Each
Description & Specifications
Specifications
Sterility Nonsterile
Length (Metric) 5.1cm
Material Polypropylene
Model D200
Packaging 10 Tipacks of 96 racked tips
Autoclavable Yes
Volume (Metric) 2 to 200μL