Please login to your online account to display your discounted pricing
Gilson™ PIPETMAN™ Tipack™ Racked Pipet Tips
Engineered to be functional, convenient and robust. Gilson™ PIPETMAN™ Tipack™ Racked Pipet Tips are available in sterilized or autoclavable packages.
Manufacturer: gilson™  F161370
 View more versions of this product
Catalog No. F161370G
  • / Pack of 600 Each
Description & Specifications
Specifications
Volume (Metric) 500 to 5000μL
Sterility Sterile
Autoclavable Yes
Model D5000
Material Polypropylene
Packaging 10 Tipacks of 96 Racked Tips
Length (Metric) 1.04cm