Please login to your online account to display your discounted pricing
Gilson™ PIPETMAN™ Tipack™ Filter Tips
Designed to meet the most stringent demands in various fields and applications such as PCR, molecular biology and cell technology. Gilson™ PIPETMAN™ Tipack™ Filter Tips are designed, developed, and manufactured in order to fit Gilson pipettes perfectly. They play a critical role in preventing aerosol-borne contamination.
Manufacturer: gilson™  F171803
 View more versions of this product
Catalog No. F171803G
  • / Pack of 960 Each
Description & Specifications
Specifications
Sterility Sterile
Length (Metric) 5.1cm
Tip Style Filter
Packaging 10 Tipacks of 96 racked tips
Model DF1200ST
Volume (Metric) 100 to 1200μL
  • Optimized geometry and pure polypropylene material ensure complete dispensing for consistent results.
  • Guarantees accuracy and precision.
  • No risk of contamination: Tips are manufactured in a clean room.
  • Gamma ray sterilized: Indicator dots used on Gilson's products undergo an irreversible color change upon sterilization.
  • Guaranteed free of DNA, RNA, DNase, RNase, ATP, Pyrogens and trace metals.
  • Full traceability from the mold to the bench guaranteed.
  • Easy identification with color-coded packaging.