Please login to your online account to display your discounted pricing
Gilson™ PIPETMAN Neo™ Starter Kits
Contains three PIPETMAN Neo™ pipets plus boxes of certified-quality PIPETMAN Tips and other useful accessories. Gilson™ PIPETMAN Neo™ Starter Kit is dedicated to laboratories which are about to renew their old set of pipets especially those performing protocols requiring intensive pipetting.
Manufacturer: gilson™  F167700
 View more versions of this product
Catalog No. F167700G
  • / Each
Description & Specifications
Specifications
Product Type Pipetteman™ Neo™ starter kit
Includes P20N, D200 and D1000 pipette, plus boxes of-quality, PIPETMAN tip D200 and D1000, Jimmy™ microtube openers, three Single™ pipette holders, a two minute inspection poster and Gilson guide to pipetteting
Includes P20N, D200 and D1000 pipette, plus boxes of-quality, PIPETMAN tip D200 and D1000, Jimmy™ microtube openers, three Single™ pipette holders, a two minute inspection poster and Gilson guide to pipetteting
Type Pipetteman™ Neo™ starter kit
Contains three pipettes conveniently packaged with useful accessories:
  • Three PIPETMAN Neo™ Pipets: P20N, P200N, and P1000N
  • Two racks of certified-quality PIPETMAN DIAMOND Tips: D200 and D1000
  • Three SINGLE Pipette holder
  • One Gilson Guide to Pipetting
  • One Two-minute inspection poster
  • Three pipette identification stickers
  • Also includes three Jimmy™ microtube openers, three Single™ pipette holders and a Gilson Guide to Pipetting