Gilson™ PIPETMAN Neo™ Multichannel Pipets
Offers an excellent compromise between comfort and target precision. Gilson™ PIPETMAN Neo™ Multichannel Pipets are designed to apply equal pressure on each piston to ensure consistent volume levels across all channels.
$1,495.83 - $1,839.31
Specifications
Product Type Multichannel Pipet
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Model Number of Channels Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Model Number of Channels Volume (Metric) Price Quantity  
F14401G Gilson™
F14401
P8x20N 8 2 to 20μL Each for $1,495.83
F14402G Gilson™
F14402
P12x20N 12 2 to 20μL Each for $1,839.31
F14403G Gilson™
F14403
P8x200N 8 20 to 200μL Each for $1,495.83
F14404G Gilson™
F14404
P12x200N 12 20 to 200μL Each for $1,839.31
Specifications
Description
  • Easy-to-remove tip ejector with autoclavable lower part
  • Internal design applies equal pressure on each piston to ensure consistent volume levels across all channels
  • PVDF handle minimizes the hand-warming effect during pipetting and is resistant to aggressive chemicals
  • Gilson Trident and unique spacer create a stepped ejection, staggering the tip ejection and thus reducing the force required to eject tips
  • Unique tip holder allows for a secure fit of most brands of tips
  • Available in 8- and 12-channel models
Order Info
Trade-in program available. Please contact your Fisher Scientific Sales representative for more details.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Multichannel Pipet