Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ L Mechanical Pipets


Combines the legendary robustness of the PIPETMAN Classic™ with a newly designed body and state-of-the-art internal mechanism to offer the superior comfort and accuracy. Gilson™ PIPETMAN™ L Mechanical Pipets are available in 8 single channel models covering the volume range of 0.2μL to 10mL.

Manufacturer: gilson™  FA10001M

 View more versions of this product

Catalog No. FA10001MG

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) D10, DL10, DF10ST, DFL10ST tip
Compatible Tips D10, DL10, DF10ST, DFL10ST tip
Accuracy ±0.024, ±0.025, ±0.027, ±0.03μL
Finger Style Ergonomic
Model P2L
For Use With (Application) Transporting a measured volume of liquid
Includes Stainless Steel tip ejector
Volume (Metric) 0.2 to 2μL
Color Orange

Mechanical pipette combines the legendary robustness of the PIPETMAN™ Classic with a newly designed body and state-of-the-art internal mechanism to offer the superior comfort and accuracy. Available in 8 single channel models covering the volume range of 0.2μL to 10mL.

Trade in program available, please contact your Fisher Scientific Sales representative for more details.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.