Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ L Fixed Pipets


New, lightweight fixed volume pipet with low forces and high reproducibility

Manufacturer: gilson™  FA10026

 View more versions of this product

Catalog No. FA10026G

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO8655, CE, GLP
Accuracy ±1.5μL
Model F250L
Color Green
Compatibility D300, DF300 tip
Compatible Tips D300, DF300 tip
For Use With (Equipment) D300, DF300 tip
Includes Plastic clip ejector
Color Green
Volume (Metric) 250μL