Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ L Fixed Pipets


New, lightweight fixed volume pipet with low forces and high reproducibility

Manufacturer: gilson™  FA10024

 View more versions of this product

Catalog No. FA10024G

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO8655, CE, GLP
Accuracy ±0.55μL
Model F100L
Color Yellow
Compatibility D200, DF100 tip
Compatible Tips D200, DF100 tip
For Use With (Equipment) D200, DF100 tip
Includes Plastic clip ejector
Color Yellow
Volume (Metric) 100μL