Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ L Fixed Pipets


New, lightweight fixed volume pipet with low forces and high reproducibility

Manufacturer: gilson™  FA10022

 View more versions of this product

Catalog No. FA10022G

  • / Each
 

Description & Specifications

Specifications

Volume (Metric) 25μL
For Use With (Equipment) D200, DF30 tip
Includes Plastic clip ejector
Compatible Tips D200, DF30 tip
Accuracy ±0.2μL
Certifications/Compliance ISO8655, CE, GLP
Color Yellow
Model F25L