Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ L Fixed Pipets


New, lightweight fixed volume pipet with low forces and high reproducibility

Manufacturer: gilson™  FA10027

 View more versions of this product

Catalog No. FA10027G

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO8655, CE, GLP
For Use With (Equipment) D1000, DF1000 tip
Model F300L
Accuracy ±2.4μL
Compatible Tips D1000, DF1000 tip
Color Blue
Volume (Metric) 300μL
Includes Plastic clip ejector