Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ G Pipets


Offers low pipetting and purge force. PIPETMAN™ G Pipet come with classic metal or plastic tip ejector.

Manufacturer: gilson™  F144054M

 View more versions of this product

Catalog No. F144054MG

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Extremely low pipetteting forces, lightweight design
Accuracy ±0.024, ±0.025, ±0.03μL
Model P2G
Color Orange
Compatibility D10, DL10, DF10ST, DFL10ST tip
Compatible Tips D10, DL10, DF10ST, DFL10ST tip
For Use With (Equipment) D10, DL10, DF10ST, DFL10ST tip
Includes Stainless Steel tip ejector
Material Stainless Steel and PVDF
Color Orange
Volume (Metric) 0.2 to 2μL