Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ G Pipets


Offers low pipetting and purge force. PIPETMAN™ G Pipet come with classic metal or plastic tip ejector.

Manufacturer: gilson™  F144066

 View more versions of this product

Catalog No. F144066G

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Extremely low pipetteting forces, lightweight design
For Use With (Equipment) D5000 tip
Includes No ejecter
Material Stainless Steel and PVDF
Color Purple
Model P5000G
Volume (Metric) 500 to 5000μL
Accuracy ±12, ±15, ±30μL
Color Purple
Compatibility D5000 tip
Compatible Tips D5000 tip