Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ G Pipets


Offers low pipetting and purge force. PIPETMAN™ G Pipet come with classic metal or plastic tip ejector.

Manufacturer: gilson™  F144059M

 View more versions of this product

Catalog No. F144059MG

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) D200, D300, DF100ST tip
Material Stainless Steel and PVDF
Compatible Tips D200, D300, DF100ST tip
For Use With (Application) Extremely low pipetteting forces, lightweight design
Accuracy ±3, ±4, ±8μL
Color Blue
Compatibility D200, D300, DF100ST tip
Includes Stainless Steel tip ejector
Model P1000G
Volume (Metric) 100 to 1000μL
Color Blue