Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ G Pipets


Offers low pipetting and purge force. PIPETMAN™ G Pipet come with classic metal or plastic tip ejector.

Manufacturer: gilson™  F144058M

 View more versions of this product

Catalog No. F144058MG

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Extremely low pipetteting forces, lightweight design
Accuracy ±0.5, ±0.8, ±1.6μL
Model P200G
Color Yellow
Compatible Tips D200, D300, DF100ST tip
For Use With (Equipment) D200, D300, DF100ST tip
Includes Stainless Steel tip ejector
Material Stainless Steel and PVDF
Color Yellow
Volume (Metric) 20 to 200μL
Compatibility D200, D300, DF100ST tip