Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ G Pipets


Offers low pipetting and purge force. PIPETMAN™ G Pipet come with classic metal or plastic tip ejector.

Manufacturer: gilson™  F144057P

 View more versions of this product

Catalog No. F144057PG

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Extremely low pipetteting forces, lightweight design
Accuracy ±0.35, ±0.4, ±0.8μL
Model P100G
Color Salmon
Compatibility D200, DF30ST tip
Compatible Tips D200, DF30ST tip
For Use With (Equipment) D200, DF30ST tip
Includes Plastic tip ejector
Material Stainless Steel and PVDF
Color Salmon
Volume (Metric) 10 to 100μL