Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ G Pipets


Offers low pipetting and purge force. PIPETMAN™ G Pipet come with classic metal or plastic tip ejector.

Manufacturer: gilson™  F144057M

 View more versions of this product

Catalog No. F144057MG

  • / Each
 

Description & Specifications

Specifications

Includes Stainless Steel tip ejector
Model P100G
Volume (Metric) 10 to 100μL
For Use With (Equipment) D200, DF30ST tip
Accuracy ±0.35, ±0.4, ±0.8μL
Color Salmon
For Use With (Application) Extremely low pipetteting forces, lightweight design
Material Stainless Steel and PVDF
Compatible Tips D200, DF30ST tip