Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ G Pipets


Offers low pipetting and purge force. PIPETMAN™ G Pipet come with classic metal or plastic tip ejector.

Manufacturer: gilson™  F144056P

 View more versions of this product

Catalog No. F144056PG

  • / Each
 

Description & Specifications

Specifications

Accuracy ±0.1, ±0.1, ±0.2μL
Compatible Tips D200, DF30ST tip
For Use With (Application) Extremely low pipetteting forces, lightweight design
For Use With (Equipment) D200, DF30ST tip
Color Yellow
Volume (Metric) 2 to 20μL
Model P20G
Color Yellow
Compatibility D200, DF30ST tip
Includes Plastic tip ejector
Material Stainless Steel and PVDF