Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ G Pipets


Offers low pipetting and purge force. PIPETMAN™ G Pipet come with classic metal or plastic tip ejector.

Manufacturer: gilson™  F144055P

 View more versions of this product

Catalog No. F144055PG

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Accuracy ±0.025, ±0.075, ±0.1μL
For Use With (Equipment) D10, DL10, DF10ST, DFL10ST tip
Material Stainless Steel and PVDF
Color Red
Model P10G
Compatible Tips D10, DL10, DF10ST, DFL10ST tip
For Use With (Application) Extremely low pipetteting forces, lightweight design
Volume (Metric) 1 to 10μL
Includes Plastic tip ejector