Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ G Pipets


Offers low pipetting and purge force. PIPETMAN™ G Pipet come with classic metal or plastic tip ejector.

Manufacturer: gilson™  F144055M

 View more versions of this product

Catalog No. F144055MG

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) D10, DL10, DF10ST, DFL10ST tip
Includes Stainless Steel tip ejector
Compatible Tips D10, DL10, DF10ST, DFL10ST tip
Model P10G
Accuracy ±0.025, ±0.075, ±0.1μL
Volume (Metric) 1 to 10μL
Color Red
Material Stainless Steel and PVDF
For Use With (Application) Extremely low pipetteting forces, lightweight design