Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN™ G Pipets


Offers low pipetting and purge force. PIPETMAN™ G Pipet come with classic metal or plastic tip ejector.

Manufacturer: gilson™  F144054P

 View more versions of this product

Catalog No. F144054PG

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

For Use With (Equipment) D10, DL10, DF10ST, DFL10ST tip
Color Orange
Model P2G
Accuracy ±0.024, ±0.025, ±0.03μL
Material Stainless Steel and PVDF
Compatible Tips D10, DL10, DF10ST, DFL10ST tip
Volume (Metric) 0.2 to 2μL
Includes Plastic tip ejector
For Use With (Application) Extremely low pipetteting forces, lightweight design