Please login to your online account to display your discounted pricing
Gilson™ PIPETMAN™ F Fixed Volume Pipettes
Dedicated fixed-volume air-displacement pipet that guarantees no risk of volume-selection errors.
Manufacturer: gilson™  F123790
 View more versions of this product
Catalog No. F123790G
  • / Each
Description & Specifications
Specifications
Includes Stainless Steel piston and PVDF handle
Accuracy ±4μL
Model F500
Warranty Lifetime
Volume (Metric) 500μL
Compatible Tips D1000, DF1000ST, DF1200ST tip
For Use With (Equipment) D1000, DF1000ST, DF1200ST tip
Certifications/Compliance GLP