Please login to your online account to display your discounted pricing
Gilson™ PIPETMAN™ F Fixed Volume Pipettes
Dedicated fixed-volume air-displacement pipet that guarantees no risk of volume-selection errors.
Manufacturer: gilson™  F123789
 View more versions of this product
Catalog No. F123789G
  • / Each
Description & Specifications
Specifications
Accuracy ±3.6μL
Certifications/Compliance GLP
For Use With (Equipment) D1000, DF1000ST, DF1200ST tip
Volume (Metric) 400μL
Compatible Tips D1000, DF1000ST, DF1200ST tip
Warranty Lifetime
Includes Stainless Steel piston and PVDF handle
Model F400
Compatibility D1000, DF1000ST, DF1200ST tip