Please login to your online account to display your discounted pricing
Gilson™ PIPETMAN™ F Fixed Volume Pipettes
Dedicated fixed-volume air-displacement pipet that guarantees no risk of volume-selection errors.
Manufacturer: gilson™  F123787
 View more versions of this product
Catalog No. F123787G
  • / Each
Description & Specifications
Specifications
Includes Stainless Steel piston and PVDF handle
Certifications/Compliance GLP
For Use With (Equipment) D1000, DF1000ST, DF1200ST tip
Volume (Metric) 250μL
Compatible Tips D1000, DF1000ST, DF1200ST tip
Warranty Lifetime
Accuracy ±3μL
Model F250