Please login to your online account to display your discounted pricing
Gilson™ PIPETMAN™ F Fixed Volume Pipettes
Dedicated fixed-volume air-displacement pipet that guarantees no risk of volume-selection errors.
Manufacturer: gilson™  F123784
 View more versions of this product
Catalog No. F123784G
  • / Each
Description & Specifications
Specifications
Model F100
Accuracy ±0.8μL
For Use With (Equipment) D200, DF100ST tip
Compatible Tips D200, DF100ST tip
Volume (Metric) 100μL
Includes Stainless Steel piston and PVDF handle
Certifications/Compliance GLP
Warranty Lifetime