Please login to your online account to display your discounted pricing
Gilson™ PIPETMAN™ F Fixed Volume Pipettes
Dedicated fixed-volume air-displacement pipet that guarantees no risk of volume-selection errors.
Manufacturer: gilson™  F123775
 View more versions of this product
Catalog No. F123775G
  • / Each
Description & Specifications
Specifications
Warranty Lifetime
For Use With (Equipment) D200, DF100ST tip
Compatible Tips D200, DF100ST tip
Certifications/Compliance GLP
Model F25
Accuracy ±0.25μL
Volume (Metric) 25μL
Includes Stainless Steel piston and PVDF handle
Compatibility D200, DF100ST tip