Please login to your online account to display your discounted pricing
Gilson™ PIPETMAN™ F Fixed Volume Pipettes
Dedicated fixed-volume air-displacement pipet that guarantees no risk of volume-selection errors.
Manufacturer: gilson™  F123772
 View more versions of this product
Catalog No. F123772G
  • / Each
Description & Specifications
Specifications
Volume (Metric) 10μL
Certifications/Compliance GLP
Accuracy ±0.1μL
Model F10
Compatibility D200, DF30ST tip
Compatible Tips D200, DF30ST tip
For Use With (Equipment) D200, DF30ST tip
Includes Stainless Steel piston and PVDF handle
Warranty Lifetime