Gilson™ PIPETMAN™ EXPERT™ Universal Fit Gel Loading Pipette Tips: Nonsterile
Flat or round tip styles are available
Manufacturer:  Gilson™ F1731182
 View more versions of this product
Catalog No. F1731182G
Specifications
Description
Tips, Pipette; Gilson; Pipetman; Gel loading; E10; Flat; Volume range: 0.1u to 10uL; Tip length: 45.72mm; O.D.: 0.17mm; Graduations at 2uL; 4 racks/Cs.
  • Color-coded tip packaging corresponds to industry standards for volume color matching to pipettes
  • Features graduations at varying volumes allowing for quick-checks when aspirating samples
  • Sterile model also available
Description & Specifications
Specifications
Gel-loading tips
Nonsterile
Flat
4.572cm
0.1 to 10μL
2μL
4/Case